EMAIL THIS PAGE TO A FRIEND
A5593 Sigma

ACGU dNTP Mix

50 ×

Purchase

Properties

concentration   50 ×
shipped in   dry ice
storage temp.   −20°C

Description

Other Notes

Contains the four dNTPs, dATP, dCTP, dGTP, and dUTP. This mix gives better contamination control for small amplicons (100-300 bp), up to 106 copies per reaction. Recommended for use with heat-labile UDG (standard UDG is not recommended for this mix).

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?