CAS Number 39389-20-3
MDL number MFCD00145841
| quality | dry |
| limit | 120 °C max. temp. |
| moisture | 52-57% |
| <3% | |
| matrix | styrene-divinylbenzene (macroreticular) |
| matrix active group | —SO3- • H+ |
| particle size | 18-23 mesh |
| 600-850 μm |
Swelling (dry to solvent saturated, %):
hexane 10-15; toluene 10-15; ethylene dichloride 15-20; ethylene acetate 30-40; 95% ethylene alcohol 60-70; water 60-70.
Strongly acidic cation exchange resin used as a heterogeneous acid catalysis; suitable for non-aqueous catalysis.
Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?