• USA Home
  • A5710 - Amberlite® MB-150 Mixed Bed Exchanger

EMAIL THIS PAGE TO A FRIEND
A5710 Sigma

Amberlite® MB-150 Mixed Bed Exchanger

gel form, 16-50 mesh (wet)

Purchase

Properties

Related Categories Cation Exchange Media, Ion Exchange Chromatography, Molecular Biology, Protein Chromatography, Proteomics More...
form   gel form
contains   ~55% moisture
feature   temperature 60 °C
particle size   16-50 mesh (wet)
capacity   0.55 meq/mL

Description

Other Notes

A mixture of 40% strongly acidic cation and 60% strongly basic anion resins.

Application

Amberlite® MB-150 Mixed Bed Exchanger can be used as a cation exchange medium in ion exchange chromatography or protein chromatography. Amberlite has been used to prepare a chelating resin, which can determine trace Pb(II) ions from solution samples, including lead ion determination in tap water samples.

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
2

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?