• USA Home
  • A5722 - Amyloid DAN Protein Fragment 1-34

EMAIL THIS PAGE TO A FRIEND
A5722 Sigma

Amyloid DAN Protein Fragment 1-34

≥75% (HPLC), solid

Synonym: Amyloid DAN Protein Precursor 277 (244-277)

  • Empirical Formula (Hill Notation) C185H270N48O51S2

  • Molecular Weight 4046.54

  •  MDL number MFCD03454661

Purchase

Description

Amino Acid Sequence

Glp-Ala-Ser-Asn-Cys-Phe-Ala-Ile-Arg-His-Phe-Glu-Asn-Lys-Phe-Ala-Val-Glu-Thr-Leu-Ile-Cys-Phe-Asn-Leu-Phe-Leu-Asn-Ser-Gln-Glu-Lys-His-Tyr

Linkage

Peptide associated with dementia.

Price and Availability


Amplified Detection

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?