• USA Home
  • A5816 - Monoclonal Anti-Antithrombin III antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
A5816 Sigma

Monoclonal Anti-Antithrombin III antibody produced in mouse

clone BL-AT III/3, ascites fluid

Purchase

Properties

Related Categories AM-AN, Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Immune Cell Signaling and Blood,
species reactivity   human
application(s)   dot blot: suitable
  indirect ELISA: suitable
  western blot: 1:200 using human antithrombin III
clone   BL-AT III/3, monoclonal
antibody form   ascites fluid
isotype   IgG1
mol wt   antigen mol wt 44-46.4 kDa (plasma)
  antigen mol wt 52.6 kDa (using purified antithrombin III)
contains   15 mM sodium azide
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... SERPINC1(462)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

human antithrombin III

Specificity

The antibody recognizes both the purified form of antithrombin III and the antithrombin III present in human plasma.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

A9522 Anti-Antithrombin III antibody produced in rabbit, fractionated antiserum, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?