• USA Home
  • A5907 - L-Arginine 4-methoxy-β-naphthylamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
A5907 Sigma

L-Arginine 4-methoxy-β-naphthylamide hydrochloride

  • CAS Number 61876-75-3

  • Empirical Formula (Hill Notation) C17H23N5O2 · HCl

  • Molecular Weight 365.86

  •  MDL number MFCD00058039

  •  PubChem Substance ID 24891034

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Fluorogenic More...
InChI Key   GWYLBAGMRDLTBD-UQKRIMTDSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?