EMAIL THIS PAGE TO A FRIEND
A5924 Sigma

Ampholine®

pH range 6.0 - 8.0

Purchase

Properties

Related Categories Carrier Ampholytes, Isoelectric Focusing and Two Dimensional Electrophoresis, Molecular Biology, Protein Electrophoresis, Proteomics More...
sterility   sterile-filtered
pH range   6.0 - 8.0
storage temp.   2-8°C

Description

Application

Ampholytes for use with polyacrylamide, agarose and dextran gels, or density gradient stabilizing media in analytical and preparative isoelectric focusing.

Other Notes

IEF carrier ampholytes

Legal Information

Ampholine is a registered trademark of GE Healthcare

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

P1897 Pharmalytes®, pH range 5.0 - 8.0

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?