• USA Home
  • A6037 - 2-Aminoethylphosphonic acid

EMAIL THIS PAGE TO A FRIEND
A6037 Sigma

2-Aminoethylphosphonic acid

Synonym: 2-AEP

Purchase

Properties

InChI Key   QQVDJLLNRSOCEL-UHFFFAOYSA-N
mp   296 °C (dec.)(lit.)

Description

Other Notes

The first carbon-phosphorus bonded compound ever reported in biological material

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

268674 2-Aminoethylphosphonic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?