EMAIL THIS PAGE TO A FRIEND
A6085 Sigma

D-(−)-Arabinose

BioXtra, ≥98%

Purchase

Properties

InChI Key   SRBFZHDQGSBBOR-ZRMNMSDTSA-N
product line   BioXtra
assay   ≥98%
impurities   ≤0.0005% Phosphorus (P)
  ≤0.1% Insoluble matter
ign. residue   ≤0.1%
mp   162-164 °C(lit.)
solubility   H2O: soluble1 M at 20 °C, clear, colorless to faintly yellow
anion traces   chloride (Cl-): ≤0.05%
  sulfate (SO42-): ≤0.05%
cation traces   Al: ≤0.0005%
  Ca: ≤0.001%
  Cu: ≤0.0005%
  Fe: ≤0.0005%
  K: ≤0.005%
  Mg: ≤0.0005%
  NH4+: ≤0.05%
  Na: ≤0.005%
  Pb: ≤0.001%
  Zn: ≤0.001%

Description

Biochem/physiol Actions

D-Arabinose is a reducing sugar. It is a pentose analog of D-ribose that is a constituent of mycobacterial cell wall arabinogalactans. It is also a substrate for D-erythroascorbic acid synthesis in yeast.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?