• USA Home
  • A6152 - β-N-Acetylglucosaminidase from human placenta

EMAIL THIS PAGE TO A FRIEND
A6152 Sigma

β-N-Acetylglucosaminidase from human placenta

ammonium sulfate suspension, 6-20 units/mg protein

Synonym: β-N-Acetyl-D-hexosaminide N-acetylhexosaminohydrolase, β-N-Acetylhexosaminidase

Purchase

Properties

Related Categories 3.2.x.x Glycosidases, 3.x.x.x Hydrolases, Application Index, Biochemicals and Reagents, Carbohydrate Hydrolysis,
form   ammonium sulfate suspension
foreign activity   α--fucosidase, and α- and β-galactosidase ≤2%
storage temp.   2-8°C
Gene Information   human ... ENGASE(64772), HEXA(3073)

Description

Biochem/physiol Actions

This enzyme, sometimes called β-N-acetylhexosaminidase, is reported to liberate terminal β-linked N-acetylglucosamine and N-acetylgalactosamine from a variety of substrates.

This enzyme, sometimes called β-N-acetylhexosaminidase, is reported to liberate terminal β-linked N-acetylglucosamine and N-acetylgalactosamine from a variety of substrates. The activity of β-N-actylglucosaminidase may be determined with the chromogenic substrate p-nitrophenyl-N-acetyl-β-D-glucosaminide. β-N-actylglucosaminidase hydrolyzes the terminal nonreducing N-acetyl-D-hexosamine residues. This enzyme contains two predominant isozymes, Hex A, a heterodimer, and Hex B, a homodimer. N-acetylglucosamine, acetamide, N-2-acetamido-2-deoyglucosylamine, N-acetylnojirimycin, and N-acetyldeoxynojirmycin are known inhibitors.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl N-acetyl-β-D-glucosaminide to p-nitrophenol and N-acetyl-D-glucosamine per min at the pH 4.25 at 25 °C.

Physical form

Suspension in 2.4 M (NH4)2SO4 containing 0.15 M NaCl and 0.1 M sodium phosphate, pH 6.0

Application

β -N-acetylglucosaminidase is a lysosomal enzyme used to hydrolyze N-acetyl-β-D-glucosaminides and N-acetyl-β-Dgalactosaminides. It is used in chemoenzymatic synthesis of oligosaccharides based on their effective transglycosylation of β-GlcNAc and β-GalNAcc. It may be a useful tool to study Alzheimer’s Disease .

Price and Availability


Amplified Detection

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Ammonium Sulfate Suspensions - Technical Note

Note that for an ammonium sulfate suspension, most of the enzyme will be in solid form. It is likely that only negligible amounts of enzyme will be in the ammonium sulfate solution.
Keywords: Enzyme activity

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

inhibitor

Product #

Description

Add to Cart

A0500 Acetamide, ~99% (GC)

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

substrate

Product #

Description

Add to Cart

M2133 4-Methylumbelliferyl N-acetyl-β-D-glucosaminide, β-N-acetylhexosaminidase substrate
N9376 4-Nitrophenyl N-acetyl-β-D-glucosaminide, ≥99% (TLC)
N4006 Naphthol AS-BI N-acetyl-β-D-glucosaminide, β-hexosaminidase substrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?