• USA Home
  • A6200 - (2R,3R)-3-Amino-2-hydroxy-4-phenylbutyric acid hydrochloride

EMAIL THIS PAGE TO A FRIEND
A6200 Sigma

(2R,3R)-3-Amino-2-hydroxy-4-phenylbutyric acid hydrochloride

  • CAS Number 128223-56-3

  • Linear Formula C10H13NO3HCl

  • Molecular Weight 231.68

  •  PubChem Substance ID 329770885

Purchase

Properties

InChI Key   OPVMPYQFOLATCK-VTLYIQCISA-N
storage temp.   −20°C

Description

Other Notes

Novel amino acid in epibestatin.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

A9075 (2S,3R)-3-Amino-2-hydroxy-4-phenylbutyric acid hydrochloride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?