• USA Home
  • A6303 - Adrenocorticotropic Hormone from porcine pituitary

EMAIL THIS PAGE TO A FRIEND
A6303 Sigma

Adrenocorticotropic Hormone from porcine pituitary

powder, 80-95 IU/mg (USP Corticotropin Ascorbic Acid Assay)

Synonym: Adrenocorticotropic Hormone Fragment 1-39, Corticotropin A

Purchase

Description

Amino Acid Sequence

Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Gly-Ala-Glu-Asp-Glu-Leu-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe

Biochem/physiol Actions

Stimulates synthesis and secretion of glucocorticoids by adrenal cortex.

Packaging

100, 250, 1000, 5000 I.U. in glass bottle

Price and Availability


Amplified Detection

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

92210 Timestrip Plus 0 °C
06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?