• USA Home
  • A6330 - Tissue Embedding Medium (Paraplast® Regular)

EMAIL THIS PAGE TO A FRIEND
A6330 Sigma

Tissue Embedding Medium (Paraplast® Regular)

Synonym: Paraffin wax

Purchase

Description

Application

Suitable for tissue infiltration. Produces minimal tissue compression and wrinkle-free sections, and cuts to 4 μm thickness with excellent ribbon continuity.

Other Notes

Double filtered paraffin wax in pellet form.

Packaging

4 x 4 lbs/case.

Legal Information

Paraplast is a registered trademark of Leica Biosystems

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
RV0350000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?