EMAIL THIS PAGE TO A FRIEND
A6332 Sigma

Antifoam 289

Purchase

Description

Application

Used in laboratories for gram positive, gram negative, yeast and fungal fermentations.

Other Notes

Antifoam 289 contains 100% active components and is a mixture of both silicone and non-silicone (organic) defoamers.

Linkage

The organic component is the same as that contained in Antifoam 204. Antifoam 289 is considered to have better dispersibility than Antifoam 204.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

A6426 Antifoam 204, mixture of organic polyether dispersions

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?