• USA Home
  • A6359 - Anti-ACTL6A (16-30) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
A6359 Sigma

Anti-ACTL6A (16-30) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-ACTL6, Anti-Actin-like 6A, Anti-Arp4, Anti-BAF53A, Anti-INO80K, Arp4

Purchase

Properties

Related Categories AC-AC, Alphabetical Index, Antibodies, Antibodies to Most Studied Epigenetic Genes, Epigenetics,
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~43 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... ACTL6A(86)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 16-30 of human ACTL6A

General description

ACTL6A is part of a family of actin-related proteins that share sequence homology with traditional actins.

Studies have reported that ACTL6A regulates the SWI/SNF complex to inhibit cell differentiation in the epidermis and thus, maintains the progenitor state. Rabbit Anti-ACTL6A binds to human ACTL6A (16-30).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Rabbit Anti-ACTL6A can be used for western blot assays at dilutions ranging from 1:500-1:2,000.

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about the Sigma Life Science and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?