• USA Home
  • A6375 - 1-Acetyl-6-chloroindoline

EMAIL THIS PAGE TO A FRIEND
A6375 Sigma

1-Acetyl-6-chloroindoline

  • CAS Number 68748-67-4

  • Empirical Formula (Hill Notation) C10H10ClNO

  • Molecular Weight 195.65

  •  MDL number MFCD00057737

  •  PubChem Substance ID 24891080

Purchase

Properties

Related Categories Building Blocks, C10, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
InChI Key   XFRSLHFDUNREPH-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?