• USA Home
  • A6450 - ω-Aminopropyl–Celite®

EMAIL THIS PAGE TO A FRIEND
A6450 Sigma

ω-Aminopropyl–Celite®

powder

Purchase

Properties

Related Categories Affinity Chromatography, Hydrophobic Interaction, Molecular Biology, Protein Chromatography, Proteomics More...
form   powder
extent of labeling   50-100 μmol per g
matrix   celite
matrix spacer   2 atoms

Description

Application

Useful for batch affinity purification; not recommended for column chromatography.

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?