• USA Home
  • A6517 - 5-(2-Aminoethylamino)-1-naphthalenesulfonic acid

EMAIL THIS PAGE TO A FRIEND
A6517 Sigma

5-(2-Aminoethylamino)-1-naphthalenesulfonic acid

suitable for fluorescence

Synonym: 1,5-EDANS, N-(Aminoethyl)-5-naphthylamine-1-sulfonic acid

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains, Luminescent Compounds/Detection, Other Fluorescent Labels,
InChI Key   SJQRQOKXQKVJGJ-UHFFFAOYSA-N
suitability   suitable for fluorescence

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?