• USA Home
  • A6532 - 3′-O-Acetylthymidine

EMAIL THIS PAGE TO A FRIEND
A6532 Sigma

3′-O-Acetylthymidine

~99%

Purchase

Properties

assay   ~99%
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?