• USA Home
  • A6542 - Anti-Rat IgG (whole molecule)−Agarose antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
A6542 Sigma

Anti-Rat IgG (whole molecule)−Agarose antibody produced in goat

IgG fraction of antiserum, saline suspension

Purchase

Properties

Related Categories Agarose, Alphabetical Index, Antibodies, Antibody Purification and Characterization, Antibody-Agaroses,
biological source   goat
antibody form   IgG fraction of antiserum
clone   polyclonal
form   saline suspension
species reactivity   rat
extent of labeling   ≥5 mg per mL
application(s)   immunoelectrophoresis: suitable
conjugate   agarose conjugate
storage temp.   2-8°C

Description

Immunogen

rat IgG

Physical form

Suspension in 0.5 M NaCl and 0.01% thimerosal

Application

Goat Anti-Rat IgG (whole molecule) has been used for immunoaffinity chromatographic applications. The antibody can also be used for immunoelectrophoresis.

Immunoprecipitation was performed in isolated primary mouse T cells using a primary antibody absorbed to agarose-linked goat anti-rat IgG antibody.

General description

IgGs are glycoprotein antibodies that modulate several immune responses. Rat IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rat IgGs are useful tools for the analysis of target proteins. Goat Anti-Rat IgG (whole molecule)-Agarose antibody binds to rat IgGs.

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?