• USA Home
  • A6560 - Agarose, low gelling temperature

EMAIL THIS PAGE TO A FRIEND
A6560 Sigma

Agarose, low gelling temperature

BioReagent, plant cell culture tested

Synonym: 2-Hydroxyethyl agarose

Purchase

Properties

Related Categories Agarose, Core Bioreagents, Gelling Agents, Molecular Biology, Plant Biotechnology,
product line   BioReagent
suitability   plant cell culture tested

Description

Analysis Note

The following is a list of properties associated with our agaroses:
Sulfate content - used as an indicator of purity, since sulfate is the major ionic group present.
Gel strength - the force that must be applied to a gel to cause it to fracture.
Gel point - the temperature at which an aqueous agarose solution forms a gel as it cools. Agarose solutions exhibit hysteresis in the liquid-to-gel transition - that is, their gel point is not the same as their melting temperature.
Electroendosmosis (EEO) - a movement of liquid through the gel. Anionic groups in an agarose gel are affixed to the matrix and cannot move, but dissociable counter cations can migrate toward the cathode in the matrix, giving rise to EEO. Since electrophoretic movement of biopolymers is usually toward the anode, EEO can disrupt separations because of internal convection.

Application

A low gelling temperature derivative with unique gelling properties. Gels form at <30 °C, remelt at temperatures in excess of 65 °C. Gels exhibit excellent clarity and are useful for the preparation of media containing heat-labile materials.
Typical working concentration: 6-12 g/L.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Agarose Product Information

CAS Number: 9012-36-6 (product A9539) CAS Number: 39346-81-1 (products A9414, A4018, A6560, A9045, A0701) Synonyms: 3,6-Anhydro-α-L-galacto-β-D-galactan; FastLane agarose; Indubiose A4; NuSieve GTG; ...
Keywords: Cell culture, Cloning, Counterimmunoelectrophoresis, Diffusion, Electrophoresis, Immunodiffusion, Immunoelectrophoresis, Indicators, Isoelectric focusing, Melting, Molecular biology, Polymerase chain reaction, Purification, Sample preparations, Separation

Related Content

Agarose Selection Guide

Agarose Selection Guide – Click on the Product Number to Purchase Specialty Agarose – Click on the Product Number to Purchase
Keywords: Cell culture, Electrophoresis, Immunodiffusion, Immunoelectrophoresis, Molecular biology, Protein electrophoresis, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?