• USA Home
  • A6626 - Nα-Acetyl-L-tyrosinamide

EMAIL THIS PAGE TO A FRIEND
A6626 Sigma

Nα-Acetyl-L-tyrosinamide

  • CAS Number 1948-71-6

  • Linear Formula 4-(HO)C6H4CH2CH(NHCOCH3)CONH2

  • Molecular Weight 222.24

  •  Beilstein Registry Number 2216288

  •  EC Number 217-756-4

  •  MDL number MFCD00002391

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

InChI Key   RJNKBEQRBIJDNM-JTQLQIEISA-N
mp   223-225 °C(lit.)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?