• USA Home
  • A6645 - 6-Acetamido-6-deoxy-α-D-glucopyranose

EMAIL THIS PAGE TO A FRIEND
A6645 Sigma

6-Acetamido-6-deoxy-α-D-glucopyranose

  • CAS Number 84064-37-9

  • Empirical Formula (Hill Notation) C8H15NO6

  • Molecular Weight 221.21

  •  PubChem Substance ID 329770900

Purchase

Properties

InChI Key   ZZYSBICZTKUVNL-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?