EMAIL THIS PAGE TO A FRIEND
A6707 Sigma
Antifoam B Emulsion
Purchase
Description
Other Notes
Antifoam A emulsion is a 30% aqueous emulsion of Antifoam A, a silicone based polymer which is the active compound. Antifoam A emulsion contains non-ionic emulsifiers.
Antifoam B Emulsion is a 10% aqueous emulsion of Antifoam A. Antifoam B emulsion contains non-ionic emulsifiers which are different from those in Antifoam A emulsion.
Price and Availability
Safety & Documentation
Safety Information
WGK Germany 
3
Documents
Certificate of Analysis
Certificate of Origin
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?