• USA Home
  • A6715 - Anti-Mouse IgG1 (heavy chain specific)−Agarose antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
A6715 Sigma

Anti-Mouse IgG1 (heavy chain specific)−Agarose antibody produced in goat

affinity isolated antibody, saline suspension

Purchase

Properties

Related Categories Agarose, Alphabetical Index, Antibodies, Antibody Purification and Characterization, Antibody-Agaroses,
biological source   goat
antibody form   affinity isolated antibody
clone   polyclonal
form   saline suspension
application(s)   immunoelectrophoresis: suitable
capacity   0.4 mg binding capacity
conjugate   agarose conjugate
storage temp.   2-8°C

Description

Immunogen

Purified mouse IgG

Physical form

Suspension in 0.5 M NaCl and 0.01% thimerosal.

Application

Mouse IgG1 (heavy chain specific)-Agarose antibody is suitable for immunoelectrophoresis. It was used for immunoprecipitation to remove excess IgG1 using using recombinant mouse lengsin and hamster vimentin.

Western blot for detection of IgM heavy chains was performed using goat anti-mouse IgG heavy chain specific.

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections. IgG1 is the most abundant of the IgG subclass
The antibody is purified by immunospecific purification and is covalently attached to cyanogen bromide activated cross-linked beaded agarose. Anti-Mouse IgG1 (heavy chain specific)−Agarose antibody is specific for mouse IgG1 when tested against purified mouse IgA, IgG (all subclasses) and IgM.

Price and Availability


Detect Your Unseen Protein
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?