• USA Home
  • A6764 - ε-Aminocaproylgalactosamine

EMAIL THIS PAGE TO A FRIEND
A6764 Sigma

ε-Aminocaproylgalactosamine

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Carbohydrate Matrices, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension
matrix   cross-linked 6% beaded agarose
matrix activation   carbonyl diimidazole
matrix attachment   amino
matrix spacer   1 atom
capacity   ~20 mg/mL binding capacity (lectin from Glycine max)(Also binds lectins from Helix pomatia, Wisteria floribunda, and Ricinus communis.)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.15 M NaCl, 0.01 M sodium phosphate, pH 6.8, containing 0.02% sodium azide

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?