• USA Home
  • A6786 - Arg-β-Ala hydrochloride

EMAIL THIS PAGE TO A FRIEND
A6786 Sigma

Arg-β-Ala hydrochloride

  • CAS Number 98957-79-0

  • Empirical Formula (Hill Notation) C9H19N5O3 · HCl

  • Molecular Weight 281.74

  •  MDL number MFCD00237775

  •  PubChem Substance ID 24891138

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   FIOGKBNTZVYASN-RGMNGODLSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?