• USA Home
  • A6984 - Anti-ADD1 (71-85) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
A6984 Sigma

Anti-ADD1 (71-85) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-ADDA, Anti-Adducin 1 (α)

Purchase

Properties

Related Categories AD-AD, Alphabetical Index, Antibodies, Primary Antibodies
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~84 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... ADD1(118)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 71-85 of human ADD1

General description

ADD1 is known to promote the differentiation of adipocytes. It is also known to regulate the genes for fatty acid metabolism.
Rabbit Anti-ADD1 (71-85) antibody binds to human ADD1 (71-85).

Adducins (α, β, γ) are cytoskeleton proteins that bind to calmondulin. ADD1 is ubiquitously expressed.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Rabbit Anti-ADD1 (71-85) antibody can be used for western blot assays at dilutions ranging from 1:500-1:2,000.

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about the Sigma Life Science and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?