EMAIL THIS PAGE TO A FRIEND
A7055 Sigma

Alprazolam-d5

Synonym: 8-Chloro-1-methyl-6-(phenyl-d5)-4H-(1,2,4)triazolo[4,3-a][1,4]benzodiazepine

Purchase

Properties

InChI Key   VREFGVBLTWBCJP-VIQYUKPQSA-N
form   neat
drug control   Home Office Schedule 4.1; psychotroperegulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
mass shift   M+5
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?