• USA Home
  • A7060 - Ala-Lys-Pro-Ser-Tyr-Hyp-Hyp-Thr-Tyr-Lys

EMAIL THIS PAGE TO A FRIEND
A7060 Sigma

Ala-Lys-Pro-Ser-Tyr-Hyp-Hyp-Thr-Tyr-Lys

≥97% (HPLC)

  • CAS Number 119106-85-3

  • Empirical Formula (Hill Notation) C55H82N12O17

  • Molecular Weight 1183.31

  •  MDL number MFCD00133051

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   RXYPJGDENAAVPR-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

Bioadhesive peptide

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?