EMAIL THIS PAGE TO A FRIEND
A7207 Sigma
Antifoam SO-25
Purchase
Description
Caution
Readily disperses in aqueous media and does not tend to coagulate or adhere to surfaces.
Other Notes
A polyol: silicone polymer
Price and Availability
Safety & Documentation
Safety Information
Flash Point(F) 
347 °F
Flash Point(C) 
175 °C
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
A8582 Antifoam SE-15, aqueous emulsion for bacterial and mammalian systems
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?