EMAIL THIS PAGE TO A FRIEND
A7342 Sigma

N-Acetyltryptamine

powder

Synonym: 3-(2-N-Acetylaminoethyl)indole

  • CAS Number 1016-47-3

  • Empirical Formula (Hill Notation) C12H14N2O

  • Molecular Weight 202.25

  •  MDL number MFCD00209910

  •  PubChem Substance ID 24278235

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Melatonins, Neurobiology, Neuroscience,
InChI Key   NVUGEQAEQJTCIX-UHFFFAOYSA-N
form   powder
solubility   alcohol: soluble
storage temp.   2-8°C
Gene Information   human ... MTNR1A(4543), MTNR1B(4544)

Description

Biochem/physiol Actions

Mixed agonist-antagonist at melatonin receptors; antioxidant.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?