• USA Home
  • A7386 - 5α-Androstan-3β-ol-16-one

EMAIL THIS PAGE TO A FRIEND
A7386 Sigma

5α-Androstan-3β-ol-16-one

≥98%

Synonym: 3β-Hydroxy-5α-androstan-16-one, Heard′s ketone

  • CAS Number 571-51-7

  • Empirical Formula (Hill Notation) C19H30O2

  • Molecular Weight 290.44

  •  MDL number MFCD00056417

  •  PubChem Substance ID 24891124

Purchase

Properties

Related Categories Androgen, Cell Biology, Cell Signaling and Neuroscience, Cytokines, Growth Factors and Hormones, Hormones,
InChI Key   WDSIJVKJXHSMCM-CJEIDTAFSA-N
assay   ≥98%
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada

Description

General description

Review of literature on Heard′s ketone

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?