• USA Home
  • A7426 - (S)-(+)-1-Aminoethylphosphonic acid

EMAIL THIS PAGE TO A FRIEND
A7426 Sigma

(S)-(+)-1-Aminoethylphosphonic acid

Synonym: D-(+)-1-Aminoethylphosphonic acid, D-Ala(P)

  • CAS Number 66068-76-6

  • Linear Formula CH3CH(NH2)P(O)(OH)2

  • Molecular Weight 125.06

  •  Beilstein Registry Number 4291033

  •  MDL number MFCD00066503

  •  PubChem Substance ID 24891205

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Phosphonic/Phosphinic Acids More...
InChI Key   UIQSKEDQPSEGAU-REOHCLBHSA-N
mp   290 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?