• USA Home
  • A7514 - Abalone acetone powder from abalone entrails

EMAIL THIS PAGE TO A FRIEND
A7514 Sigma

Abalone acetone powder from abalone entrails

crude

Purchase

Properties

Related Categories Acetone Powders, Biochemicals and Reagents, Proteins and Derivatives More...
grade   crude
storage temp.   −20°C

Description

Application

Crude source of β-glucuronidase and sulfatase

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?