• USA Home
  • A7561 - N-Acylmannosamine 1-Dehydrogenase from Pseudomonas sp.

EMAIL THIS PAGE TO A FRIEND
A7561 Sigma

N-Acylmannosamine 1-Dehydrogenase from Pseudomonas sp.

recombinant, expressed in E. coli, lyophilized powder, ≥45 units/mg protein

Synonym: N-Acyl-D-mannosamine: NAD 1-Oxidoreductase

Purchase

Properties

Related Categories 1.1.x.x Acting on hydroxyl groups, 1.x.x.x Oxidoreductases, Biochemicals and Reagents, Carbohydrate Metabolism, Cell Biology,
recombinant   expressed in E. coli
form   lyophilized powder
mol wt   mol wt ~120 kDa (gel filtration)
solubility   0.1 M Tris-HCl, pH 8.2: 5 mg/mL
storage temp.   −20°C

Description

Biochem/physiol Actions

Enzymatically converts N-acetyl-D-mannosamine to N-acetyl-D-mannosaminolactone.

Unit Definition

One unit will oxidize 1.0 μmole of N-acetyl-D-mannosamine to N-acetyl-D-mannosaminolactone per minute at pH 8.2 at 37 °C in the presence of NAD.

Physical form

Powder also contains bovine albumin and sucrose.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?