• USA Home
  • A7702 - Monoclonal Anti-Aspartate antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
A7702 Sigma

Monoclonal Anti-Aspartate antibody produced in mouse

clone ASP-1, ascites fluid

Purchase

Properties

Related Categories AS-ATG, Alphabetical Index, Antibodies, Antibodies for Neurotransmission, Antibodies to Neurotransmitters,
biological source   mouse
antibody form   ascites fluid
clone   ASP-1, monoclonal
contains   15 mM sodium azide
application(s)   indirect ELISA: 1:10,000 using in situ prepared L-aspartate-glutaraldehyde-BSA conjugate for coating
isotype   IgG2a
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Immunogen

L-aspartic acid (Asp) conjugated to KLH.

Specificity

The antibody reacts specifically with L-aspartate (L-aspartic acid), a major neurotransmitter in the central nervous system, when conjugated with glutaraldehyde. When using the antibody at the recommended working dilution in indirect and competitive ELISA, weak cross-reaction may be obtained with D-aspartate, L-glutamate, D-glutamate, L-glutamine, L-asparagine, β-alanine, glycine, 5-aminovaleric acid, γ-aminobutyric acid (GABA), Gly-Asp, Gly-Gly and Asp-Glu.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?