• USA Home
  • A7710 - 1,4,6-Androstatriene-3,17-dione

EMAIL THIS PAGE TO A FRIEND
A7710 Sigma

1,4,6-Androstatriene-3,17-dione

≥95% (HPLC), powder

Purchase

Properties

Related Categories Androgen, Cell Biology, Cell Signaling and Neuroscience, Cytokines, Growth Factors and Hormones, Hormones,
assay   ≥95% (HPLC)
form   powder
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
solubility   methanol: soluble50 mg/mL

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BV8082250

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?