• USA Home
  • A7751 - trans-Aconitic anhydride

EMAIL THIS PAGE TO A FRIEND
A7751 Sigma

trans-Aconitic anhydride

90-95%

  • CAS Number 14556-16-2

  • Empirical Formula (Hill Notation) C6H4O5

  • Molecular Weight 156.09

  •  MDL number MFCD00238750

Purchase

Properties

InChI Key   UNCRKEZQEKKGQX-UHFFFAOYSA-N
assay   90-95%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

217808 cis-Aconitic anhydride, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?