• USA Home
  • A7909 - 4-(2-aminoethylamido)succinyl avidin

EMAIL THIS PAGE TO A FRIEND
A7909 Sigma

4-(2-aminoethylamido)succinyl avidin

Synonym: Avidin, 4-(2-aminoethylamido)succinyl

Purchase

Properties

composition   Protein, ~90% biuret
storage temp.   −20°C

Description

Physical form

Lyophilized powder containing sodium citrate

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?