• USA Home
  • A7916 - 2-Acetamido-2-deoxy-6-O-(β-D-galactopyranosyl)-D-glucopyranose

EMAIL THIS PAGE TO A FRIEND
A7916 Sigma

2-Acetamido-2-deoxy-6-O-(β-D-galactopyranosyl)-D-glucopyranose

Synonym: β-D-Gal-(1→6)-D-GlcNAc

  • CAS Number 50787-10-5

  • Empirical Formula (Hill Notation) C14H25NO11

  • Molecular Weight 383.35

  •  MDL number MFCD00057922

  •  PubChem Substance ID 24891292

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates A-C, Disaccharide More...
InChI Key   FGYNENQLWILFLQ-UHFFFAOYSA-N
storage temp.   −20°C

Description

Biochem/physiol Actions

Inhibitor of lectins

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?