• USA Home
  • A7924 - Albumin, bovine-α-D-mannopyranosylphenyl isothiocyanate-biotin labeled

EMAIL THIS PAGE TO A FRIEND
A7924 Sigma

Albumin, bovine-α-D-mannopyranosylphenyl isothiocyanate-biotin labeled

lyophilized powder

Synonym: α-D-Mannosylated-biotin-albumin

Purchase

Properties

form   lyophilized powder
extent of labeling   15-20 mol α-D-mannopyranose per mol albumin
  ~3 mol biotin per mol albumin
impurities   salt, free
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?