EMAIL THIS PAGE TO A FRIEND
A8074 Sigma

ADA

BioXtra, ≥98% (titration)

Synonym: N-(2-Acetamido)iminodiacetic acid, N-(Carbamoylmethyl)iminodiacetic acid

Purchase

Properties

Related Categories BioXtra Buffers, Biochemicals and Reagents, Biological Buffers, Buffers A to Z, Good’s Buffers More...
InChI Key   QZTKDVCDBIDYMD-UHFFFAOYSA-N
Quality Level   ELITE
product line   BioXtra
assay   ≥98% (titration)
impurities   ≤0.005% Phosphorus (P)
  ≤0.1% Insoluble matter
useful pH range   6.0 - 7.2
pKa (25 °C)   6.6
mp   219 °C (dec.)(lit.)
solubility   1 M NaOH: soluble0.05 M, clear, colorless
anion traces   chloride (Cl-): ≤0.05%
  sulfate (SO42-): ≤0.05%
cation traces   Al: ≤0.0005%
  Ca: ≤0.0005%
  Cu: ≤0.0005%
  Fe: ≤0.0005%
  K: ≤0.005%
  Mg: ≤0.0005%
  NH4+: ≤0.05%
  Pb: ≤0.001%
  Zn: ≤0.0005%

Description

Application

Biological buffer component with sodium hydroxide and sodium chloride (pH 5.67-7.57, useful pH range 6.4-7.4). Used to prepare immobilized pH gradients.

Price and Availability


Interactive Buffer Table
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Buffer Reference Center

Citric Acid – Na2HPO4 Buffer Solutions, pH approx. 2.6–7.61 Citric acid monohydrate, C6H8O7 • H2O, M. wt. 210.14; 0.1M-solution contains 21.01 g/l. Na2HPO4, M. wt. 141.98; 0.2M-solution contains 28.4...
Keywords: Biological Buffers, Buffers

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?