• USA Home
  • A8134 - L-Arginine Decarboxylase from Escherichia coli

EMAIL THIS PAGE TO A FRIEND
A8134 Sigma

L-Arginine Decarboxylase from Escherichia coli

Type IV, partially purified powder, 200-400 units/mg protein

Synonym: L-Arginine carboxylyase, ADC, Arginine decarboxylase

Purchase

Properties

type   Type IV
form   partially purified powder
composition   Protein, ~5% Lowry
storage temp.   −20°C

Description

Unit Definition

One unit will release 1.0 μmole of CO2 from L-arginine per min at pH 5.2 at 37°C.

Physical form

Partially purified powder containing succinate buffer salts and sucrose

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

A5381 L-Arginine Decarboxylase from Escherichia coli, Type II, lyophilized powder, 5-15 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?