• USA Home
  • A8162 - ω-Aminooctyl Matrix: Cross-linked 4% beaded agarose

EMAIL THIS PAGE TO A FRIEND
A8162 Sigma

ω-Aminooctyl Matrix: Cross-linked 4% beaded agarose

saline suspension

Purchase

Properties

form   saline suspension
extent of labeling   4-8 μmol per mL
matrix activation   epoxy
matrix attachment   amino of 1,8-diaminooctane
matrix spacer   12 atoms
capacity   4-8 mg/mL binding capacity (bovine serum albumin)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?