• USA Home
  • A8175 - Ammonium metavanadate

EMAIL THIS PAGE TO A FRIEND
A8175 Sigma

Ammonium metavanadate

cell culture tested, powder

Synonym: Ammonium vanadate(V)

Purchase

Properties

InChI Key   YBVKNHXQSRDWAA-UHFFFAOYSA-M
form   powder
color   white
density   2.32 g/cm3 at 25 °C(lit.)
suitability   cell culture tested

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2859 6.1 / PGII
WGK Germany 
3
RTECS 
YW0875000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?