EMAIL THIS PAGE TO A FRIEND
A8215 Sigma

L-Asparagine-UL-14C

≥95% (Radiochemical Purity, HPLC), aqueous ethanol solution

Purchase

Properties

InChI Key   CKLJMWTZIZZHCS-BGTGBAJNSA-N
assay   ≥95% (Radiochemical Purity, HPLC)
form   aqueous ethanol solution
extent of labeling   120-240 mCi per mmol
storage temp.   2-8°C

Description

Packaging

Combi-vial

Physical form

Aqueous solution containing 2% ethanol

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?