• USA Home
  • A8219 - Sodium acetate-UL-14C

EMAIL THIS PAGE TO A FRIEND
A8219 Sigma

Sodium acetate-UL-14C

≥95% (HPLC), aqueous solution

Synonym: Acetic acid-UL-14C sodium salt

  • Linear Formula 14C2H3O2Na

  • Molecular Weight 82.03

  •  MDL number MFCD00189465

Purchase

Properties

assay   ≥95% (HPLC)
form   aqueous solution
extent of labeling   40-60 mCi per mmol
storage temp.   2-8°C

Description

Packaging

Combi-vial

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?