• USA Home
  • A8223 - [Leu31]-Amyloid β-protein fragment 1-40 trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
A8223 Sigma

[Leu31]-Amyloid β-protein fragment 1-40 trifluoroacetate salt

≥95% (HPLC), lyophilized powder

Purchase

Properties

assay   ≥95% (HPLC)
form   lyophilized powder
mol wt   calculated mol wt 4331
composition   Protein Content, ~80%
solubility   H2O: soluble
storage temp.   −20°C

Description

Amino Acid Sequence

Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Leu-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val

Other Notes

Substituted form of Aβ [1-40]

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?