• USA Home
  • A8276 - [Sar1, Leu8]-Angiotensin II acetate salt hydrate

EMAIL THIS PAGE TO A FRIEND
A8276 Sigma

[Sar1, Leu8]-Angiotensin II acetate salt hydrate

≥97% (HPLC)

  • Empirical Formula (Hill Notation) C46H73N13O10 · xC2H4O2 · yH2O

  • Molecular Weight 968.15 (anhydrous free base basis)

  •  MDL number MFCD00133977

Purchase

Properties

Related Categories Angiotensins, Cell Biology, Cell Signaling and Neuroscience, Peptides and Proteins, Peptides for Cell Biology More...
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Sar-Arg-Val-Tyr-Ile-His-Pro-Leu

Biochem/physiol Actions

Angiotensin II antagonist

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?